This product is manufactured in our Grand Island, US facility and is intended for sale in North America, Latin America, and the Asia/Pacific region—higher freight charges will apply to orders shipped elsewhere. An equivalent product, manufactured in Paisley, Scotland, is recommended for our customers in Europe, Africa, and the Middle East. Click to view.
Gibco™

PBS, pH 7.2 This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  20012050

 Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco® PBS formulations for a range of cell culture applications.

This PBS is manufactured as follows:

Without
• Calcium
• Magnesium
• Phenol Red

The complete formulation is available.

cGMP Manufacturing and Quality System
Gibco® PBS is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical Gibco® PBS product made in our Scotland facility (70011-036). This facility is also registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
For In Vitro Diagnostic Use.

Specifications

Concentrated: 1 X
Inorganic Salts: No Calcium, No Magnesium
pH: pH 7.2
Form: Liquid
Classification: Animal Origin-Free
Osmolality: 280 - 320 mOsm/kg
Phenol Red Indicator: No Phenol Red
Product Line: Gibco®
Product Size: 10 x 500 mL
Sodium Pyruvate Additive: No Sodium Pyruvate
pH Range: 7.1 - 7.3
Green Features: Sustainable packaging

Contents & storage

Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture

Media formulations

Documents

Manuals & protocols

Recommended products