Gibco™

Ham's F-12K (Kaighn's) Medium This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  21127022

 Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ham's F-12K (Kaighn's) Medium is a modification of Ham's F-12 Nutrient Mixture. Ham's F-12K (Kaighn's) Medium was developed for primary human hepatocytes, as well as for some rat and chicken liver cells in a reduced serum environment.

This F-12K (Kaighn's) Medium is manufactured as follows:
WithWithout
• L-glutamine• HEPES
• Phenol Red

The complete formulation is available.

Using Ham's F-12K
Ham's F-12K (Kaighn's) Medium contains many components not found in traditional basal media, such as putrescine, thymidine, hypoxanthine, zinc, and higher levels of all amino acids and sodium pyruvate. These additions allow the medium to be supplemented with very low levels of serum or defined components, for some cell types. Ham's F-12K (Kaighn's) Medium contains no proteins or growth factors, and is therefore often supplemented with growth factors and Fetal Bovine Serum (FBS). The FBS concentration must be optimized for each cell line. Ham's F-12K (Kaighn's) Medium uses a sodium bicarbonate buffer system (2.5 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Dual-site cGMP manufacturing
For supply chain continuity, we manufacture Ham's F-12K (Kaighn's) Medium at two separate facilities, located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP-manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Glucose: Low Glucose
Glutamine: L-Glutamine
HEPES Buffer: No HEPES
Phenol Red Indicator: Phenol Red
Product Line: Gibco®
Product Size: 500 mL
Sodium Pyruvate Additive: Sodium Pyruvate
Volume (Metric): 500 ml
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Media formulations

Documents

Manuals & protocols

Help center

Cell Culture & Transfection Learning Center

Access cell culture and transfection educational resources for better experiment planning and execution.

Cell Culture & Transfection Support Centers

Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.

Recommended products