Neomycin Sulfate

Catalog number: 21810031

Gibco™  Related applications: Insect Cell Culture | Mammalian Cell Culture | Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Gibco® Neomycin is produced by Streptomyces fradiae. Polymyxin B Sulfate acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria. Gibco® Neomycin Sulfate is used for the prevention of bacterial contamination of cell cultures. This antibiotic is highly active against gram-negative bacteria. The recommended working concentration is 50 µg/ml. We offer a variety of antibiotics and antimycotics for cell culture applications.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Validated Application: Bacterial Selection
Agent: Neomycin
Form: Powder
Reagent Type: Antibiotic (Selective)
Product Size: 100 g
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture

Documents

Manuals & protocols

Recommended products