Gibco™

Glasgow's MEM, powder

Catalog number:  22100093

 Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Glasgow's Minimum Essential Medium (GMEM) was originally developed by Ian McPherson and Michael Stoker as a modification of Eagle's medium. It was utilized to study the genetic factors which affected cell competence. Glasgow's MEM was developed for use with kidney cells lines, such as BHK-21.
This MEM is modified as follows:
WithWithout
• L-glutamine• Tryptose phosphate broth
• Phenol Red• Sodium Bicarbonate
• HEPES

The complete formulation is available.

Glasgow's MEM is unique from other media as it contains twice the concentration of amino acids and vitamins than the original Basal Medium Eagle, and is used without serum. Glasgow's MEM was originally formulated with 10% tryptose phosphate broth.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.

cGMP Manufacturing
Gibco® Glasgow's MEM is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Glasgow's MEM contains no proteins, lipids, or growth factors. Therefore, Glasgow's MEM requires supplementation, commonly with 10% tryptose phosphate broth. Glasgow's MEM uses a sodium bicarbonate buffer system (2.75 g /) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Powder forms of Gibco® cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Specifications

Form: Powder
Glucose: High Glucose
Glutamine: L-Glutamine
HEPES Buffer: No HEPES
Phenol Red Indicator: Phenol Red
Product Line: Gibco®
Product Size: 50 L
Sodium Pyruvate Additive: No Sodium Pyruvate
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8°C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture

Media formulations

Documents

Manuals & protocols

Recommended products