LB Agar, powder (Lennox L agar)

Catalog number:  22700025

Invitrogen™  Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Formulation per one liter:
10 g SELECT Peptone 140
5 g SELECT Yeast Extract
5 g sodium chloride
12 g SELECT Agar
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Final Product: Agar Plates
Preparation Method: Autoclave
Form: Powder
Target Organism Class: E. coli
Product Size: 500 g
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Media formulations

Documents

Manuals & protocols

Recommended products