Proteinase K (Fungal)

Catalog number:  25530031

Invitrogen™  Related applications: Chromatin Biology | DNA Extraction | RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins. Proteinase K remains active:

• Over a wide pH range—optimal activity between 6.5 and 9.5
• Under denaturing conditions—e.g., in the presence of SDS or urea
• In the presence of metal chelating agents—e.g., EDTA
• At comparatively high temperatures—optimum digestion temperature is 65°C

Applications
Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and quality testing
Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit definition
One mAnson unit is described as that amount of enzyme that liberates 1 µmole of Folin-positive amino acid within 1 min at 37°C using hemoglobin as a substrate.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration: >40 U/mg
Form: Powder
Product Size: 1 g
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

Contents & storage

One tube Proteinase K
Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products