Spectra™ Multicolor Low Range Protein Ladder

Catalog number:  26628

Thermo Scientific™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Thermo Scientific Spectra Multicolor Low Range Protein Ladder is a mixture of six proteins and synthetic peptides (1.7 to 40kDa) for use as size standards for peptides and small proteins in gel electrophoresis and Western blotting.

Features of Spectra Multicolor Low Range Protein Ladder:

Size range—six proteins and peptides spanning 1.7 to 40kDa
Multicolor—three different colors for unambiguous band-size assignment
Ready-to-use—supplied in a loading buffer for direct loading on gels; no need to boil
Sharp bands—color-coded bands of similar intensity for easy visualization
Quality tested—each lot evaluated by SDS-PAGE and Western blotting
Membrane-compatible—colored bands transfer to membranes for Western blotting

• Dye-stained proteins in 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol.

• Monitoring of protein migration during SDS-PAGE
• Verifying Western transfer efficiency
• Approximate sizing of proteins on SDS-polyacrylamide gels and Western blots
• Locating a protein of interest for excision from an unstained preparative gel

This prestained protein MW marker is designed for monitoring the progress of SDS-polyacrylamide gel electrophoresis, for assessing transfer efficiency onto PVDF, nylon and nitrocellulose membranes, and for estimating the approximate size of separated proteins that have been made visible with gel stains or Western blot detection reagents. Three different chromophores (blue, orange, green) are bound to the different component proteins, producing a brightly colored ladder with an easy-to-remember pattern. The Spectra Multicolor High Range Protein Ladder is ready to use: no heating, further dilution or addition of a reducing agent is required.
For Research Use Only. Not for use in diagnostic procedures.


Stain Type: Prestained
System: SDS-PAGE
Product Line: Spectra™
Product Size: 250 µL

Contents & storage

Upon receipt store at -20°C. Product is shipped with an ice pack.


Manuals & protocols

Recommended products