Medium 199, Earle's Salts, powder

Catalog number: 31100019

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco® Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This M199 is modified as follows:

WithWithout
• L-glutamine• HEPES
• Phenol Red• Sodium bicarbonate

The complete formulation is available.

Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. Medium 199 is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2.

In Vitro Diagnostic
Gibco® Medium 199 is manufactured in compliance with the requirements of ISO 13485: 2003 for the design, development, manufacture, and distribution of liquid and powder tissue culture media, sera, and reagents for cell culture research, industrial bioprocessing, and related markets.

Dual-Site cGMP Manufacturing
Gibco® Medium 199 is manufactured in a cGMP New York facility that is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 . For supply chain continuity, we offer an identical Gibco® Medium 199 product made in the Scotland facility (31100-019). This second cGMP manufacturing location is also registered with the FDA as a medical device manufacturer and is certified to ISO 13485. Medium 199 contains no proteins, lipids, or growth factors. Medium 199 uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH. Powder forms of Gibco® cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Specifications

Glutamine: L-Glutamine
Phenol Red Indicator: Phenol Red
Form: Powder
HEPES Buffer: No HEPES
Sodium Pyruvate Additive: No Sodium Pyruvate
Glucose: Low Glucose
Product Line: Gibco®
Product Size: 50 L
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture

Media formulations

Documents

Manuals & protocols

Recommended products