This product is manufactured in our Grand Island, US facility and is intended for sale in North America, Latin America, and the Asia/Pacific region—higher freight charges will apply to orders shipped elsewhere. An equivalent product, manufactured in Paisley, Scotland, is recommended for our customers in Europe, Africa, and the Middle East. Click to view.

Ham's F-12 Nutrient Mix, GlutaMAX™ Supplement

Catalog number: 31765035

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free single-cell plating of Chinese Hamster Ovary (CHO) cells. F-12 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including chondrocytes and rat prostate epithelial cells. We offer a variety of Gibco® F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This Ham's F-12 is modified as follows:

With
• GlutaMAX™
• Phenol Red

The complete formulation is available.

Compared to other basal media, F-12 contains a wider variety of components, including zinc, putrescine, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-12 has led to a variety of improved formulations. Gibco® F-12 with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

cGMP Manufacturing and Quality System
For supply chain continuity, we manufacture Gibco® F-12 at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.

Gibco® F-12 contains no proteins or growth factors. Therefore, F-12 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-12 uses a sodium bicarbonate buffer system (1.176 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
For In Vitro Diagnostic Use.

Specifications

Form: Liquid
Glutamine: GlutaMAX™-I
HEPES Buffer: No HEPES
Product Size: 500 mL
Volume (Metric): 500 ml
Phenol Red Indicator: Phenol Red
Sodium Pyruvate Additive: Sodium Pyruvate
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Documents

Manuals & protocols

Recommended products