Related applications: Mammalian Cell Culture
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
With |
• GlutaMAX™ |
• Phenol Red |
Form: | Liquid |
---|---|
Glutamine: | GlutaMAX™-I |
HEPES Buffer: | No HEPES |
Phenol Red Indicator: | Phenol Red |
Product Line: | Gibco®, GlutaMAX™ |
Product Size: | 10 x 500 mL |
Sodium Pyruvate Additive: | Sodium Pyruvate |
Volume (Metric): | 500 ml |
Green Features: | Sustainable packaging |
Shipping Condition: | Room Temperature |
Catalog number: | 11875119 |
Size | 10 x 500 mL |
Price | USD 209.00 |
Add to favorites | |
Catalog number: | 14190250 |
Size | 10 x 500 mL |
Price | USD 179.00 |
Add to favorites | |
Catalog number: | 35050061 |
Size | 100 mL |
Price | USD 34.26 |
Add to favorites | |