Dynabeads® M-270 Carboxylic Acid, for OEM and industrial use only

Catalog number: 34300D

 Related applications: Protein Assays and Analysis | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Dynabeads® M-270 Carboxylic Acid are monosized hydrophilic 2.8-µm superparamagnetic beads, composed of highly crosslinked polystyrene. Carboxylic acid groups on the surface allow for covalent amide bond formation to nucleic acids, proteins?peptides, and other ligands via primary amino- or sulphydryl groups, without requirement for protein blocking. Activation through carbodiimide is required and alternative reactive groups can be introduced by using commercially available cross-linkers.

Dynabeads® magnetic beads are widely used as a solid-phase for presenting antibodies or antigens in automated clinical immunoassay systems, where the combination of specific antigens or antibodies and the superparamagnetic properties of the beads provide rapid reaction kinetics both in the coating process, separation, and during washing of the analyte.

The product meets the reputable Dynal high standards with respect to reproducibility and automation ability, and drives reliability for your assays.

Benefits:

• Efficient and reproducible immobilization of antibodies (or other ligands)
• Easy and reproducible handling in manufacturing
• Rapid separation and washing
• Reproducible behavior in automation without mixing requirements, with fast and efficient washing procedures
• Fast coupling without need for protein blocking
• Generally low background and high signal-to-noise ratio

Applications
Ideally suited for magnetic bead-based immunoassays requiring a low background signal.

Note:
This particular product format is for large volume customers, available on an OEM basis. This product is also available in smaller volumes (14306D and 14305D).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Surface Functionality: Carboxylic Acid, Hydrophilic
Concentration: 100 mg⁄ml
Product Line: Dynabeads®
Product Size: 10 mL
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols