Invitrogen™

Dynabeads™ SILANE Viral NA Kit

Catalog number:  37011D

 Related applications: Diagnostic Development | RNA Extraction | Viral RNA⁄DNA Purification

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

For sensitive, highly predictable and consistent extraction and isolation of viral DNA and RNA from human serum/plasma samples, following a simple separation protocol. The 1 µm Dynabeads have an optimized silica-like surface chemistry, and a high specific surface area, providing efficient kinetics and a high sensitivity. Their uniform size and surface area ensure reproducible results.

A higher sensitivity and improved performance is achieved with Dynabeads® compared to magnetic particles from alternative suppliers. Other particles often have a random size range distribution, surface area and binding capacity. This could compromise the reproducibility of your results. Dynabeads® hold reputable Dynal high standards with respect to within- and between-lot reproducibility and automation ability.

Advantages:

• Isolates viral DNA and viral RNA
• High sensitivity, allowing for a low detection limit
• Effective isolation of nucleic acids even from challenging viruses (e.g. HBV)
• Can detect HBV DNA in a 200 µl serum sample containing only 40 infectious units (using 2 mg (50 µl) Dynabeads®)
• Quicker than spin-columns
• Cost-effective and performs to the same level as spin-columns
• Automation-friendly (slow sedimentation rate + high magnetic mobility)

Applications:
• Isolation of viral nucleic acids (DNA/RNA)
• From human serum/plasma samples
• Well suited for automated assays
• Separate kit available for the isolation of genomic DNA (Cat. no. 370-12D)

Additional Info:
Validation and customization (bead, buffer, protocol or format) can be made available on a custom OEM basis. For further information, please contact us at: ivd@invitrogen.com
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): Liquid Samples (e.g. Serum), Viral Samples
Sample Type (Specific): Plasma, Serum
Downstream Application: Northern Blotting, Southern Blotting
High Throughput Compatibility: High Throughput-Compatible
Isolation Technology: Magnetic Bead
Product Line: DYNAL®, Dynabeads®
Product Size: 96 preps
Shipping Condition: Room Temperature

Contents & storage

The kit contains: Dynabeads® MyOne™ SILANE (5 ml, 40 mg/ml, see Cat. no. 370-02D for further characteristics), binding/washing/elution buffers and a protocol optimized for isolation of viral DNA/RNA. The kit content is sufficient for 96 isolations.
Store at: 2-8°C.

Documents

Manuals & protocols

Recommended products