FAK Antibody (ZF002)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Specifications

Reactivity: Human, Mouse, Rat
Validated Application: Western Blot
Host Species: Mouse
Clonality: Monoclonal
Clone ID: ZF002
Target Gene: Ptk2
Gene ID: 14083, 25614
Label or Dye: Unconjugated
Purification: Affinity Purified
Product Size: 100 µg
Protein Family: Kinases & Inhibitors, Extracellular Matrix & Cell Adhesion
Research Category: Signal Transduction, Extracellular Matrix & Cell Adhesion
Shipping Condition: Wet Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products