Thermo Scientific™

HAAKE™ CaBER™ 1 Capillary Breakup Extensional Rheometer

 Related applications: Rheometers & Viscometers

Easily quantify extensional properties of semi-solids and fluids with the Thermo Scientific™ HAAKE™ CaBER™ 1 Capillary Breakup Extensional Rheometer.

The HAAKE CaBER 1 Capillary Breakup Extensional Rheometer is the only commercially available extensional rheometer to quantify elongational properties of fluids or semi-solids. The thinning and break-up of a fluid filament that is analyzed with the HAAKE CaBER 1 Capillary Breakup Extensional Rheometer provides valuable information on a material's physical properties that rotational rheometers simply cannot provide.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

The HAAKE CaBER 1 Capillary Breakup Extensional Rheometer Features:

  • Computer-controlled operation
  • Easy sample loading and cleaning
  • Class 1 laser micrometer
  • Linear motor drive with variable speed
  • Automatic repeated testing
  • Closed temperature-controlled sample chamber
  • Exchangeable geometries
  • User defined strains
  • Small sample volume (< 0.2mL)
Recommended for:
  • Adhesives (solvent loss or gain)
  • Food products
    • Stringiness/strand formation
    • Time to breakup
    • Relaxation of doughs
    • Elastic instabilities
    • Mouth feeling
  • Consumer goods
    • Filling of bottles
    • Time to breakup
    • Solvent loss
    • Processability
  • Industrial resins
    • Relaxation time spectrum
    • Constitutive modeling
    • Spinnability
  • Printer inks (stringiness/strand formation)

Food Solutions through Rheology & Extrusion

Learn about instruments to help you create, process and monitor food products to have ideal texture, shape, mouthfeel, processability, shelf life and more.



Material characterization webinars

In our hour-long webinars, you will be provided with in-depth knowledge on rheology, viscometry, compounding, or extrusion in general or how you can develop and optimize your innovative applications, products or workflows.
Watch now ›

Documents

Manuals & protocols