Interested in a bulk or custom version of this product? Request pricing.

HA Recombinant Influenza A, subtype H1N1 (A/England/195/2009) Protein, His Tag

Catalog number: 40005V08H25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Hemagglutinin (HA), recombinant viral protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of the influenza A H1N1 (A/England/195/2009 (H1N1)) hemagglutinin (ACR15621.1) (Met 1-Gln 529, HA1+HA2, uncleved) fused to a C-terminal polyhistidine tag.

Activity: This recombinant protein has not been tested.

Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Microbiology, Virology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: HA
Gene Symbol: HA
Molecular Weight: 59 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Cell Structure⁄Organelle Proteins
Protein Form: Recombinant, Truncated
Protein Name: Hemagglutinin
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Influenza
Tag Position: C-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols