Trichloroacetic Acid Dichloromethane, Deblocking Solution

Catalog number: 400236

Applied Biosystems™  Related applications: Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Contains 450 mL Trichloroacetic Acid Dichloromethane Deblocking Solution.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name: Trichloroacetic Acid Dichloromethane
For Use With (Equipment): 3900 DNA Synthesizer
Form: Liquid
Product Size: 450 mL
Shipping Condition: Room Temperature

Contents & storage

Store in a cool, dry, and well-ventilated location.

Documents

Manuals & protocols