Gibco™

NCTC-135 Medium This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  41350026

 Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

NCTC-135 medium is a chemically defined medium that was originally developed at the National Cancer Institute (Bethesda, MD) and formulated for the culture of mouse Strain L sublines, NCTC clone 929 (L929 cells) in a serum-free environment. Gibco® NCTC-135 medium has also been used to grow many cells, including HeLa, Ehrlich-Lettre Ascites strain E, NCTC clone 1469, NCTC 4206, mouse C3H, BS-C-1, and hybridoma cell lines.
This NCTC-135 is modified as follows:
WithWithout
• L-glutamine• HEPES
• Phenol Red
• Coenzymes

The complete formulation is available.

Gibco® NCTC-135 is a modification of NCTC 109 medium that replaced L-cysteine with L-cystine, which reduced toxicity to some cells. NCTC-135 medium is rich in nucleosides contains several unique components to eliminate the need for serum, including coenzymes, glucosamine, reducing agents and additional vitamins.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco® NCTC-135 medium is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

NCTC-135 medium contains no proteins, lipids, or growth factors. NCTC-135 medium was designed for use without serum for L929 cells, but typically requires supplementation of a growth factor cocktail or Fetal Bovine Serum (FBS) for other cells. NCTC-135 medium uses a sodium bicarbonate buffer system (2.2 g / L) and therefore requires a 5-10% CO2 environment to maintain physiological pH. Some cell lines might require gradual adaptation to a serum-free medium.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Glutamine: L-Glutamine
Product Line: Gibco®
Product Size: 500 mL
Volume (Metric): 500 ml
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Media formulations

Documents

Manuals & protocols

Recommended products