Gibco™

Ham's F-10 Nutrient Mix, GlutaMAX™ Supplement This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  41550021

 Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells. F-10 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including COS-7, primary rat astrocytes, and rat prostate epithelial cells.

This F-10 is modified as follows:

WithWithout
• GlutaMAX™• HEPES
• Phenol Red

The complete formulation is available.

Compared to other basal media, F-10 contains a wider variety of components, including zinc, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-10 has led to a variety of improved formulations, such as Ham's F-12 Nutrient Mixture (F-12).

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco® F-10 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Gibco® F-10 contains no proteins or growth factors. Therefore, F-10 often requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-10 uses a sodium bicarbonate buffer system (1.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Specifications

Classification: Animal Origin-Free
Concentrated: 1 X
Culture Environment: CO2
Form: Liquid
Glutamine: GlutaMAX™-I
HEPES Buffer: No HEPES
Osmolality: 275 - 315 mOsm/kg
Phenol Red Indicator: Phenol Red
Product Line: Gibco®, GlutaMAX™
Product Size: 500 mL
Sodium Bicarbonate Buffer: Sodium Bicarbonate
Sodium Pyruvate Additive: Sodium Pyruvate
Supplementation Required: Serum
Volume (Metric): 500 ml
pH Range: 7.0 - 7.4
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8° C. Protect from light.
Shipping conditions: Ambient.
Shelf life: 12 months from date of manufacture.

Documents

Manuals & protocols

Recommended products