RPMI 1640 Medium, HEPES, no glutamine This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  42401018

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of Gibco® RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This RPMI 1640 is modified as follows:
WithWithout
• Phenol Red• L-glutamine
• HEPES

The complete formulation is available.

Gibco® RPMI 1640 is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12, and PABA which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations.

cGMP Manufacturing and Quality System
Gibco® RPMI 1640 is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

RPMI 1640 contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
For In Vitro Diagnostic Use.

Specifications

Glutamine: No Glutamine
Phenol Red Indicator: Phenol Red
Form: Liquid
HEPES Buffer: HEPES
Serum Supplementation: Standard Serum Supplementation
Sodium Pyruvate Additive: No Sodium Pyruvate
Classification: Animal Origin-Free
Concentrated: 1 X
Culture Environment: CO2
Osmolality: 270 - 310 mOsm/kg
Product Line: Gibco®
Product Size: 500 mL
Sodium Bicarbonate Buffer: Sodium Bicarbonate
Supplementation Required: Serum
Volume (Metric): 500 ml
pH Range: 7.0 - 7.4
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8° C. Protect from light.
Shipping conditions: Ambient.
Shelf life: 12 months from date of manufacture.

Documents

Manuals & protocols

Recommended products