TC-100 Insect Medium, powder

Catalog number:  43000033

Gibco™  Related applications: Insect Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

TC 100 Insect Medium was originally designed for the growth of insect cells, including Spodeptera frugiperda (Sf9), and the expression of recombinant Autographa Californica Nuclear Polyhedrosis Virus (rAcNPV) using the Baculovirus Expression Vector System (BEVS). We offer a variety of Gibco® Media options for a range of insect cell culture applications.

This IPL-41 Insect Medium is made as follows:

WithWithout
• L-glutamine • Sodium bicarbonate
• Tryptose phosphate broth

The complete formulation is available. TC 100 Insect Medium is rich in amino acids and uses tryptose as an additional sugar source.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

cGMP Manufacturing and Quality System
Gibco® TC 100 Insect Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Gibco® TC 100 Insect Medium is commonly supplemented with 10% Fetal Bovine Serum (FBS) before use. TC 100 Insect Medium requires ambient CO2 in order to maintain physiological pH. The powder form of TC 100 Insect Medium requires the addition of 350 mg/L sodium bicarbonate before use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line: Sf21, Sf9
Cell Type: Insect Cells
Form: Powder
Product Line: Gibco®
Product Size: 10 L
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8°C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture

Media formulations

Documents

Manuals & protocols