Six Paq GeneAmp® dNTP Blend 10mM

Catalog number: 4303441

Applied Biosystems™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

GeneAmp® dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. GeneAmp® dNTPs with dUTP can be used to generate dU-containing PCR products in a carry-over prevention strategy in conjunction with AmpErase ® Uracil N-glycosylase (UNG).

• dNTPs have high purity, making them ideal for use in PCR reactions or other applications.
• dNTPs with dUTP are optimized for use in carry-over prevention.
• Guaranteed for PCR performance when used with GeneAmp® PCR core reagents and GeneAmp® instrument systems.
• Premixed dNTP blends are available in various concentrations, offering ease of use.

Each vial consists of a 1mL of a 10mM blend consisting of a soln of 2.5mM each of dATP, dCTP, dGTP, dTTP. This part number includes the following part numbers that may or may not be sold separately: N8080260

The PCR process is covered by patents owned by Roche Molecular Systems, Inc. and F. Hoffmann-La Roche Ltd.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration: 10 mM
Product Line: GeneAmp®
Product Size: 6 x 1 mL

Contents & storage

Store at -20°C in a constant temperature freezer.

Documents

Manuals & protocols