Applied Biosystems™

GeneAmp™ dNTP Blend (100 mM)

Catalog number:  4303443

 Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

GeneAmp® dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH.

• dNTPs have high purity, making them ideal for use in PCR reactions or other applications.
• Guaranteed for PCR performance when used with GeneAmp® PCR core reagents and GeneAmp® instrument systems.
• Premixed dNTP blends are available in various concentrations, offering ease of use.

Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP). Sufficient for 2000 PCR amplifications, 50 µL each. Each kit contains 12 x 1 mL vials.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration: 100 mM
Product Line: GeneAmp®
Product Size: 12 x 1 mL

Contents & storage

Store at -20°C in a constant temperature freezer.

Documents

Manuals & protocols