Related applications: Real Time PCR (qPCR)
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
Calibration Type: | Spectral Calibration |
---|---|
Detection Method: | Fluorescent |
For Use With (Equipment): | 7700 System |
Label or Dye: | FAM™, JOE™, NED™, ROX™, SYBR® Green, TAMRA™, TMR (Tetramethylrhodamine), VIC® |
Product Size: | 8 tubes |
Shipping Condition: | Dry Ice |