Need help? Find end-point PCR and RT-related tips, troubleshooting help, and technical resources within our new PCR and cDNA Synthesis Support Center.
Applied Biosystems™

MicroAmp™ Clear Adhesive Film

Catalog number:  4306311

 Related applications: Automated Nucleic Acid Purification | PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Applied Biosystems® MicroAmp® Clear Adhesive Film creates a secure seal across the entire microplate to prevent evaporation and well-to-well contamination.

Features:

• Adhesive provides tight seal over wells to prevent leaks and evaporation.
• Easy to apply and remove.
• Available as Clear (economical and sticky) or Optical (ready for real-time PCR and non-tacky).

Reduce Leaks, Evaporation, and Well-to-Well Contamination
This clear adhesive film seals your samples securely into the wells of your microplates, reducing leaks, evaporation, and contamination among sample wells.

Easy to Use
Simply peel the backing away from the adhesive film and apply firmly across the top of a microplate. Once sealed, you can safely stack the microplates without worrying about contamination. Use the MicroAmp® Adhesive Seal Film Applicator to speed the process of sealing multiple microplates.


Note: See user's manual or package insert for limited label license, and trademark information. For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Clear
For Use With (Equipment): GeneAmp™ 9700, ProFlex™ PCR System, SimpliAmp™ Thermal Cycler, Veriti® Dx Fast Thermal Cycler, Veriti® Dx Thermal Cycler, Veriti® Fast Thermal Cycler, Veriti® Thermal Cycler
Plate Compatibility: 384-Well Plates, 96-Well Plates
Product Line: MicroAmp®
Product Size: 100 films
Shipping Condition: Room Temperature
Type: Film, Seal, or Applicator

Contents & storage

Contains: 100 films
Storage: Room temperature

Documents

Manuals & protocols

Recommended products