AmpFLSTR™ Identifiler™ PCR Amplification Kit

Catalog number: 4322288

Applied Biosystems™  Related applications: Forensics

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The AmpFLSTR™ Identifiler™ PCR Amplification Kit is the single most discriminating and widely used STR based kit for human identification applications from Applied Biosystems™. The Identifiler kit amplifies 15 loci and Amelogenin in a single tube and provides loci consistent with major world-wide STR databasing standards.

Features of the The AmpFLSTR™ Identifiler™ PCR Amplification Kit:

• Fifteen STR (short tandem repeat) loci and Amelogenin co-amplified in a single tube.
• Loci consistent with all major worldwide STR standards.
• Degenerate unlabeled primer addresses mutations observed in a population of Chamorros and Filipinos from Guam
• Includes 13 core loci required for sample entry in CODIS

Complete Validated Solution
Applied Biosystems offers a complete solution for your human identification needs. Our system includes DNA quantification kits, a range of AmpFLSTR™ STR kits, instrumentation, software, and consumables. The Identifiler kit is part of the most comprehensively validated systems for STR-based Human Identification. All reagents are developed in accordance with guidelines from leading worldwide forensic organizations such as SWGDAM and DAB.

Five Dye Technology Allows Enhanced Throughput
Identifiler utilizes the most advanced 5-dye technology to allow for the multiplex of more loci in a single lane or injection helping to increase your throughput and to simplify the workflow needed to obtain probative results. Our PET™ and LIZ™ -dyes were specifically designed to expand the spectral range of the 4-dye system. We have expanded the spectral range to 660 nm allowing for maximum color separation with minimal spectral overlap. As with all of our products, the five-dye technology is validated for use with our latest instruments and software as a complete system.

Experience Matters™
When you partner with us you gain access to the most extensive Human Identification resource network found within the industry. Our dedicated team of industry leading experts includes Research and Development scientists with extensive forensic product development experience, skilled Field Applications Support Scientists with over 100 years of combined forensic experience, and dedicated Field Service Engineers that provide on-site technical assistance with instrumentation.

Note: For Forensic or Paternity Use Only.
For Forensic or Paternity Use Only.

Specifications

Amplicon Size: <360 bp
For Use With (Equipment): 310 Genetic Analyzer, 3100-Avant™ Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, GeneAmp® 9600, GeneAmp™ 9700
Label or Dye: FAM™, LIZ™, NED™, PET™, VIC®
Matrix and Size Standards: GeneScan™ 500 LIZ® Size Standard, Matrix Standard Set DS-33
Polymerase: AmpliTaq Gold® DNA Polymerase
Product Line: AmpFLSTR™, Identifiler®
Product Size: 200 reactions
Reaction Format: SuperMix or Master Mix
Sample Type (Specific): DNA (Genomic), Forensic Sample
Sample Volume: 25 µL⁄reaction
Technique: STR (Short Tandem Repeat) Analysis
Shipping Condition: Dry Ice

Contents & storage

• AmpFLSTR™ Identifiler PCR Reaction Mix
• AmpFLSTR™ Identifiler Primer Set
• AmpFLSTR™ Control DNA 9947A
Store at 2 to 8°C.

• AmpliTaq™ Gold DNA Polymerase
Store at -15 to -25 °C.

• AmpFLSTR™ Identifiler™ Allelic Ladder
Store at -15 to -25°C upon receipt, 2 to 8°C after initial use

Documents

Manuals & protocols

Recommended products