This Applied Biosystems® easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™, JOE™, NED™,ROX™, SYBR™ Green, TAMRA™, TET™, and VIC® dyes on the 7900HT Sequence Detection System. The Sequence Detection Systems Spectral 384-Well Calibration kit includes two optical 384-well reaction plates: one preloaded and sealed background plate, and one preloaded and sealed spectral calibration plate with eight separate dye standards. • 384-well optical reaction plates preloaded with dyes (FAM™, JOE™, NED™, ROX™, SYBR™ Green, TAMRA™, TET™, and VIC®) are provided for quick and easy calibration of the ABI PRISM® 7900HT Sequence Detection System • Easy-to-use and provides accurate discrimination of the fluorescence signal of individual dyes in the reaction mixture • Includes the Passive Reference I dye standard for signal normalization in all real-time and endpoint reactions Spend Your Time on Research—Not Instrument Calibration This kit's easy-to-use, preloaded, 96-well optical reaction plates enable users to calibrate the 384-well block of the 7900HT Sequence Detection System quickly and easily. Insert the reaction plates during system installation, and the system automatically collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibrated Dye
FAM, JOE, NED, ROX, TAMRA™, TMR (Tetramethylrhodamine), VIC, SYBR™ Green
For Use With (Equipment)
AB 7900 HT 384-well System
Detection Method
Fluorescent
Product Type
Spectral Calibration Kit
For Use With (Application)
Real Time PCR (qPCR)
Shipping Condition
Dry Ice
Description
384-well Plate
Quantity
2 x 384-well Plates
Calibration Type
Spectral Calibration
Contents & Storage
Includes one Background and one Spectral Calibration plate with eight dye standards plates (FAM™, JOE™, NED™, ROX™, SYBR™ Green, TAMRA™, TET™, and VIC™).
Store at -15 to 30°C. Protect from light.
Expiration date: 12 months from the date of delivery