This product is now part of a food safety range that includes Thermo Scientific™ culture media, molecular solutions, and other products for your complete microbiological workflow. In US, Canada and Europe, order this product from Thermo Fisher Scientific microbiology. >> Order now

TaqMan® GMO Maize 35S Detection Kit

Catalog number: 4327690

Applied Biosystems™

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This kit contains reagents needed to reliably detect the presence of genetically modified organism (GMO)-specific DNA sequences in seed, grain, processed foods and their ingredients.
• Universal assay detects and quantifies all approved genetically modified organisms (GMOs) in Europe and the vast majority of GMOs approved in other countries.
• Fluorogenic 5' nuclease assay using TaqMan® probes provides accurate and reproducible quantitative results.
• Label license conveys the PCR service rights for GMO testing when used in conjunction with an Authorized Thermal Cycler, so no additional royalties are payable.
• Comprehensive controls provide high confidence in results.
• Innovative GMO quantitation software allows users to quickly and easily determine the percentage of GMO content.
• Offers the complete solution including sample preparation, instruments, reagents, analysis software, technical support, instrument service, and GMO expertise.

The Complete Solution
The TaqMan® GMO Detection and Quantitation Kit is the first of a full product line of GMO assays in development that can detect the presence of GMO-specific DNA sequences in seed, grain, and processed foods and their ingredients. The TaqMan® GMO kit is part of a highly sophisticated but user-friendly system that includes DNA sample preparation, automated PCR amplification and signal detection, data analysis, and quantitation software, thus eliminating the need for personnel trained in molecular techniques.

Reliable TaqMan® Chemistry
The system uses the patented fluorogenic 5' nuclease method for multiplex detection and quantitation, the latest innovation in real-time PCR technology. It targets the Cauliflower Mosaic Virus 35S promoter, a GMO-specific sequence present in all GMO soy and maize events approved for use in food by the European Union, and in the vast majority of GMO events approved by other countries. Both the European Union and Japan view this method as the premier technology for quantitating GMO content in food. Other TaqMan® kits are available for the detection and quantitation of food-borne pathogens, including Salmonella and E. coli 0157:H7.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Primer-Probe Detection
For Use With (Equipment): 7300 System, 7500 System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
Hot Start: Built-In Hot Start
Number of Reactions: 100 Tests
PCR Method: qPCR
Passive Reference Dye: ROX (Pre-mixed)
Polymerase: AmpliTaq Gold® DNA Polymerase
Primer-Probe Compatibility: TaqMan®
Product Line: Applied Biosystems™, TaqMan™
Product Size: 1 kit
Reaction Speed: Standard
Sample Type (General): Plant Samples
Sample Type (Specific): Food, Seed
Target Organism Class: Maize, Genetically Modified
Shipping Condition: Dry Ice

Contents & storage

Each kit contains TaqMan GMO Mix, AmpliTaq Gold® DNA Polymerase, positive and negative controls. Each kit contains enough regents for 100 assays.

Store all components at -25 to -15°C.

Documents

Manuals & protocols