Applied Biosystems™

Premium USB Type A-B Cable, 3 meters (for 7500 & 7500 Fast Systems)

Catalog number:  4328260

 Related applications: Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is a spare part for the 7300, 7500, and 7500 Fast Sequence Detection Systems.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 7500 Fast System, 7500 System
Product Size: 1 cable
Shipping Condition: Room Temperature
Type: USB Cable

Contents & storage

Store at room temperature.

Documents

Manuals & protocols