Related applications: Genotyping & Genomic Profiling | Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
Block Format: | Interchangeable |
---|---|
Calibrated Dye: | FAM™, JOE™, NED™, ROX™, SYBR® Green I, TAMRA™, TET™, VIC™ |
Capacity: | Up to 36 x 96-well plates with optional automation accessory, Up to 84 x 384-well plates (with optional automation accessory and 384-well block) |
Detection Method: | Primer-Probe Detection, SYBR |
Dimensions: | 72 cm (W) x 84 cm (D) x 64 cm (H) (with drawer open) |
Dynamic Range: | Up to 9 logs of linear dynamic range |
For Use With (Equipment): | 7900HT Fast System |
Format: | 96-well plate |
High Throughput Compatibility: | High Throughput-Compatible, Multiplexing |
Optics: | Extended-life 488 nm argon-ion laser excitation source, No filters (CCD acts as spectrograph with continuous detection 500 -650 nm) |
Passive Reference Dye: | No ROX, Other Passive Reference Dye, ROX (Pre-mixed), ROX (Separate Tube) |
Product Size: | 1 instrument |
Reaction Speed: | Standard |
Reaction Volume Range: | 20-100 µl |
Run Time: | <2 hrs (standard mode) |
Sample Ramp Rate: | Fast mode: ± 3.0°C⁄sec, Standard mode: ± 1.6°C⁄sec |
Sensitivity: | Down to 10 copies |
Temperature Accuracy: | ±0.25°C of display temperature |
Temperature Range (Metric): | 4-100°C |
Temperature Uniformity: | ±0.5°C |
Thermal Cycling System: | Peltier-Based System |
Throughput: | ≥10,000 endpoint PCR rxns per hour (384-well block and automation accessory), ≥5,000 real-time PCR rxns per day (384-well block and automation accessory) |
Weight: | 114 kg (250 lbs) with automation accessory, 82 kg (180 lbs) without automation accessory |