SNaPshot® Primer Focus® Kit

Catalog number: 4329538

Applied Biosystems™  Related applications: DNA Sequencing | Fragment Analysis | Genotyping & Genomic Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The SNaPshot® PrimerFocus® kit allows rapid assessment of potential SNP oligonucleotides. You can preview all potential single-base extension products and calculate the mobility rate for each allele. After assessing this data, you can determine the optimal combination of SNP markers for multiplexing. After the multiplex format has been determined, you can use the reference data to establish markers and bin sets quickly in the GeneMapper® software, thereby reducing the time required to define and edit bins manually.

Used With:
SNaPshot® Multiplex Kits

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Analysis Software: GeneMapper® Software
Detection Method: Capillary Electrophoresis
For Use With (Equipment): 310 Genetic Analyzer, 3100-Avant™ Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer
Format: Tube(s)
Genotyping Target: SNPs (Known)
High Throughput Compatibility: Multiplexing
Hot Start: No
Polymerase: AmpliTaq® DNA Polymerase
Primer-Probe Compatibility: Unlabeled Oligos
Product Line: Applied Biosystems™, Primer Focus®, SNaPshot®
Product Size: 100 reactions
Sample Type (General): DNA
Technique: Primer Extension, SNP Genotyping (Fragment Analysis-Based)
Shipping Condition: Dry Ice

Contents & storage

This kit contains the reagents (ddNTP mix, enzyme, buffer and co-factor) and a control primer.
Store frozen at -15 to -25°C

Documents

Manuals & protocols