Human RPLPO (Large Ribosomal Protein) Endogenous Control (FAM™/MGB probe, non-primer limited) This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: 4333761T

Applied Biosystems™  Related applications: Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Applied Biosystems® Human RPLPO (large ribosomal protein) Endogenous Control (FAM™ ⁄ MGB Probe, Non-Primer Limited) is intended as an endogenous control. It allows relative gene expression quantification in cDNA samples when used with other gene expression assays. Probe is labeled with 6FAM™ dye - MGB and the primers are not limited. Can be used for singleplex PCR reactions. Endogenous control is to be used with Inventoried and Non-Inventoried TaqMan® Gene Expression Assays, Custom TaqMan® Gene Expression Assays, and⁄or Custom TaqMan® Primers and Probes.

Assay Details:

Gene Symbol: RPLPO
RefSeq: NM_053275.3
Probe Exon Location:3
Amplicon Size: 105

TaqMan® Endogenous Controls

Eliminate months of assay design, formulation, and testing by using TaqMan® Endogenous Controls as your controls to quantify gene expression. This convenient collection of pre-designed probe and primer sets enables you to normalize the amount of sample RNA or DNA in a reaction.

Complete Solution for Quantitative Gene Expression

Having a hard time deciding what controls to use to quantify gene expression — even with detailed information on biological systmes? Now, with TaqMan® Endogenous Controls, you can avoid all the trial-and-error of selecting controls for most common human, mouse, rat, and eukaryotic genes.

Simple to Use

All components of the TaqMan® Endogenous Controls are QC tested, formulated into a single 20X mix, and functionally tested. The controls are not only simple to use, but they are also fully compatible with universal conditions for two-step RT-PCR. Just add TaqMan® Universal PCR Master Mix (with or without AmpErase® UNG) and your cDNA sample to generate sensitive, reproducible, and truly quantitative gene expression data on Applied Biosystems instruments including the Applied Biosystems 7900HT, 7300, 7500 Real-Time PCR Systems, and the 7000 and 7700 Sequence Detection Systems.

Flexible Offering

We build each endogenous control using our proven 5' nuclease chemistry. For maximum flexibility, you can choose between two different reporter dyes and two quenchers:
• A FAM™ dye-labeled TaqMan® MGB probe (250nM, final concentration) and two unlabeled PCR primers (900nM each)
• A VIC® dye-labeled TaqMan® MGB probe (250nM, final concentration) and two unlabeled PCR primers (150nM each y primer limited)
• A VIC® dye-labeled TAMRA™ probe (250nM, final concentration) and two unlabeled PCR primers (150nM each y primer limited)

Choosing the Right Endogenous Control

Endogenous controls can normalize the expression levels of target genes by correcting differences in the amount of cDNA that is loaded into PCR reaction wells. For best results, verify that the endogenous control is consistently expressed in the sample set to be tested. Endogenous control expression must be uniform across all samples in the study. For multiplexing, ensure that the gene expression level of the endogenous control is greater than that of the target.

Multiplex vs. Singleplex PCR

All TaqMan® Endogenous Controls that contain probes labeled with the VIC® reporter dye are primer limited. This allows multiplexing of TaqMan® Endogenous Controls with target gene expression assays, provided that the control gene is more abundantly expressed than the target gene. All TaqMan® Endogenous Controls that contain probes labeled with the FAM™ reporter dye are not primer limited and are not intended for multiplexing .

For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: Tube(s)
Species: Human
Includes: Control Primer-Probe Set(s)
Gene Symbol: RPLPO
Target Gene: RPLPO
Label or Dye: FAM™
Product Size: 125 reactions
Amplicon Size: 105
Accession Number: NM_053275.3
Green Features: Less waste, Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Package includes 1 tube containing 20X mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 °C to -25 °C

Documents

Manuals & protocols