BigDye™ Terminator v3.1 Cycle Sequencing Kit

Catalog number:  4337456

Applied Biosystems™  Related applications: DNA Sequencing | Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The BigDye® Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry is ideal for de novo sequencing, resequencing, and finishing with PCR product, Plasmid, Fosmid, and BAC templates.
• Improve the quality of your results for a wide range of sequencing applications.
• Optimized for long read lengths.
• Better dye mobility characteristics
• Improved performance reading through GT rich regions.
• Get longer, higher-quality reads with more uniform peak heights and optimal signal balance.
• Enhance your productivity and reduce costs.

Get More Uniform Peak Heights, Improved Accuracy
With BigDye® Terminator v3.1 kit superior chemistry, you generate data with uniform peak heights and optimized signal balance. This results in longer, higher-quality reads and more accurate base assignments for heterozygote and mutation detection.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 310 Genetic Analyzer, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, GeneAmp™ 9700, Veriti® Thermal Cycler
Recommended Applications: AT-Rich Sequencing (>65%), Comparative Sequencing (Germline Mutations - 50:50), De Novo Sequencing - Low to Mid Throughput, GC-Rich Sequencing (>65%), GT-Rich or Difficult Template Sequencing, Long-Read Sequencing, Mixed-Base Detection, Resequencing, Standard Sequencing
Format: 12-strip wells, 384-well plate, 8-well strips, 96-well plate
Product Line: BigDye™
Product Size: 1,000 reactions
Technique: Fluorescent Dye Terminator Sequencing
Template Compatibility: BAC DNA, Fosmids, Genomic DNA (Bacterial), Lambda DNA, PCR Amplicons (50:50 Heterozygotes), Plasmid DNA (≤15Kb), Rolling Circle Amplified Products, Single Stranded DNA

Contents & storage

• 10 tubes of BigDye® Terminator v3.1 Ready Reaction Mix
• M13 (-21) Primer
• pGEM®-3Zf(+) double-stranded DNA Control Template
• 1 × 12 ml tube of 5X Sequencing Buffer (may be stored at 4°C)

Store kit at -15°C to -25°C and avoid repeated freeze-thaw cycles.

Documents

Manuals & protocols

Recommended products