Applied Biosystems™

AmpliTaq Gold™ DNA Polymerase, LD (Low DNA) with Gold Buffer and MgCl2

Catalog number:  4338856

 Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Applied Biosystems® AmpliTaq Gold® DNA Polymerase, LD (Low DNA) is a highly purified version of AmpliTaq Gold® DNA Polymerase that significantly reduces contaminating bacterial DNA sequences commonly present in recombinant protein preparations.

• Manufacturing process results in highly purified enzyme with lower levels of bacterial DNA
• Minimizes non-target, false positive PCR products when amplifying bacterial sequences
• Chemically modified hot-start technique eliminates biological contamination
• GeneAmp® 10X PCR Gold Buffer promotes faster enzyme activation and robust PCR amplification
• Enhanced specificity with time-release PCR is ideal for low-copy target amplification
• AmpliTaq Gold® DNA Polymerase, LD improves both yield and success rate for target amplification

Reliable Results
This highly purified enzyme preparation gives you more confidence in your results and makes advanced PCR techniques quick and easy. When combined with the supplied GeneAmp® 10X PCR Gold Buffer, AmpliTaq® Gold DNA Polymerase, LD provides a flexible hot-start system that is suitable for all PCR applications that require the elimination of bacterial DNA contamination.

Note:
See user's manual or package insert for limited label license and trademark information. For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: AmpliTaq Gold® DNA Polymerase
Hot Start: Built-In Hot Start
Fidelity (vs. Taq): 1 X
GC-Rich PCR Performance: Low
Reaction Format: Separate Components
Product Overhang: 3'-A
Reaction Speed: Standard
Final Product Size(s): 5 kb or less
Exonuclease Activity: 5' - 3'
Product Line: AmpliTaq Gold™
Product Size: 250 units
Purity or Quality Grade: LD (Low DNA)

Contents & storage

Contains AmpliTaq Gold® DNA Polymerase, LD (250 units), GeneAmp® 10X PCR Gold Buffer, and MgCl2 Solution. Store at –15 to –30°C.

Documents

Manuals & protocols

Recommended products