Phosphoramidite Diluent

Catalog number: 4340858

Applied Biosystems™  Related applications: Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Contains 4 amidite diuent (anhydrous acetonitrile) filled Trap-Pak™ bags.

The anhydrous acetonitrile comes in Trap-Pak™ bags. Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to efficiently adsorb water. Use Trap-Pak™ bags to reduce and maintain a low water content in acetonitrile and tetrazole (activator) solutions.

• Extensive testing and quality controls assure high quality reagents that meet our high expectations and specifications.
• Reagents and supplies are use-tested and optimized for the 3900 system for quality nucleic acid synthesis.

Tested
Our high-performance reagents begin with raw materials that must conform to our strict specifications. Once the reagents are manufactured and packaged, extensive testing is performed to assure you that all liquids are free of interfering particulate matter; reagent concentrations are within our specifications, and the performance specification meets at least 98% stepwise coupling efficiency.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name: Phosphoramidite
For Use With (Equipment): 3900 DNA Synthesizer
Form: Liquid
Product Line: Applied Biosystems™
Product Size: 4 x 100 mL

Contents & storage

Topfill consisting of four each of amidite diluent 100 ml Part Number 401445
Store in a cool, dry, and well-ventilated location.

Documents

Manuals & protocols