Tetrazole Acetonitrile

Catalog number: 4340859

Applied Biosystems™  Related applications: Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This tetrazole acetonitrile solution comes in Trap-Pak™ bags. Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to efficiently adsorb water. Use Trap-Pak™ bags to reduce and maintain a low water content in acetonitrile and tetrazole (activator) solutions.

• Extensive testing and quality controls assure high quality reagents that meet our high expectations and specifications.
• Reagents and supplies are use-tested and optimized for the 3900 system for quality nucleic acid synthesis.

Tested
Our high-performance reagents begin with raw materials that must conform to our strict specifications. Once the reagents are manufactured and packaged, extensive testing is performed to assure you that all liquids are free of interfering particulate matter; reagent concentrations are within our specifications, and the performance specification meets at least 98% stepwise coupling efficiency.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name: Tetrazole Acetonitrile
For Use With (Equipment): 3900 DNA Synthesizer
Form: Liquid
Product Size: 4 x 450 mL

Contents & storage

(topfill consisting of four each of 450 ml tetrazole acetonitrile Part Number 401173
Store in a cool, dry, and well-ventilated location.

Documents

Manuals & protocols