Tetrazole Acetonitrile

Catalog number: 4340864

Applied Biosystems™  Related applications: Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is a four-pack of 180 mL Tetrazole Acetonitrile solution.

This Tetrazole Acetonitrile solution comes in a convenient large volume for ABI 394 Nucleic Acid synthesizers, resulting in extended hands-free DNA synthesis. With these larger bottles attached your synthesizer can produce up to 40 average primer-length oligonucleotides with no bottle changes, reducing attended operation time by 50%.

• Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
• Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing. oligonucleotide synthesis yields of full-length product.
• Reagent kits are available for your convenience, and save you time and money.

Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name: Tetrazole Acetonitrile
For Use With (Equipment): 3900 DNA Synthesizer
Form: Liquid
Product Line: Applied Biosystems™
Product Size: 4 x 180 mL

Contents & storage

Topfill consisting of four bottles of Tetrazole Acetonitrile 180 ml, Part Number 400606
Store in a cool, dry, and well-ventilated location, appropriate for flammable lliquids.

Documents

Manuals & protocols