Applied Biosystems™

1- Methylimidazole/THF, Capping Formula B

Catalog number: 4340866
Applied Biosystems™

1- Methylimidazole/THF, Capping Formula B

Catalog number: 4340866
Catalog Number
4340866
Unit Size
4 x 180 mL
Price (USD)
Price: 504.00
Your price:
Availability
-
Quantity
Catalog Number
Unit Size
180 mL
Price (USD)
Price: 540.00
Your price:
Availability
-
Quantity
Catalog Number
Unit Size
450 mL
Price (USD)
Price: 904.00
Your price:
Availability
-
Quantity
Catalog Number
Unit Size
4 x 450 mL
Price (USD)
Price: 868.00
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43408664 x 180 mL
Price: 504.00
Your price:
-
400785180 mL
Price: 540.00
Your price:
-
401175450 mL
Price: 904.00
Your price:
-
43408614 x 450 mL
Price: 868.00
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Contains 4 CapB filled Trap-Pak™ bags.

Cap B is a mix of NMI (N-methylimidazole) and THF (tetrahydrofuran).

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Liquid
For Use With (Equipment)
3900 DNA Synthesizer
Product Type
1-methylimidazole/tetrahydrofuran
Quantity
4 x 180 mL
Reagent Type
Solvent or General Reagent

Contents & Storage

4 × 180 ml

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text