Applied Biosystems™

Tempus™ Blood RNA Tube

Catalog number:  4342792

 Related applications: Automated Nucleic Acid Purification | RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Tempus™ Blood RNA Tubes are for stabilization and isolation of total RNA from whole blood for gene expression analysis. Advantages of Tempus™ Blood RNA Tubes:

• Immediately lyse whole blood cells and stabilize RNA in a single step—no RBC lysis or proteinase K treatments required
• Freeze global gene expression profiles for up to five days at room temperature; greater than seven days at 4°C

How it works
Each Tempus™ Blood RNA Tube contains 6 mL Stabilizing Reagent. When the blood (3 mL) is drawn into the tube and mixed with this reagent, lysis occurs almost immediately. The stabilizing reagent inactivates cellular RNases and selectively precipitates RNA; genomic DNA (gDNA) while proteins remain in solution. Blood drawn into the Tempus™ Blood RNA Tube is stable for up to five days at room temperature, or minimally seven days at 4°C.

Accessory product
When the Tempus™ Blood RNA Tubes are used with the Tempus™ 12-Port RNA Isolation Kit (Cat. No. 4378672), RNAs with greater stability and higher yields of RNA are produced (see figure).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: Tempus™
Product Size: 50 tubes
Shipping Condition: Room Temperature
Tube Type: Sample Tube

Contents & storage

Store unused tubes at room temperature.

Documents

Manuals & protocols

Recommended products