Product 4345287 will be available until supplies run out. It is now replaced by Cat. no. 100052500, which is an equivalent Halogen Lamp.
Applied Biosystems™

Halogen Lamp for 7300, 7500, and 7500 Fast Systems

Catalog number:  4345287

 Related applications: Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This halogen lamp is an accessory for 7300, 7500, and 7500 Fast Systems.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 7300 System, 7500 Fast System, 7500 System
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Lamp

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products