Applied Biosystems™

MicroSEQ™ D2 rDNA Fungal PCR Kit

Catalog number:  4349153

 Related applications: Microbial Identification & Detection | Microbial Protein Production

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This kit is the PCR component of the MicroSEQ® Fungal Identification System, which provides an easy-to-use DNA sequence-based method to identify most fungi. A protocol and Quick Reference Card are not included.
• Sequence-based measurement ensures you get reproducible results in less than 24 hours.
• Growth requirement-independent measurement enables you to identify fungi using objective criteria.
• Universal domain-level primers eliminate up-front optimization.
• MicroSEQ® ID Analysis Software ensures accurate species identification.
• Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
• Requires minimal training; same work flow as the MicroSEQ® Full Gene 16S rDNA Bacterial Identification Kit.

Specific and Easy to Use
Based on phylogenetic analysis of the D2 region of large-subunit ribosomal RNA gene sequences, this easy-to-use kit enables you to identify even the most difficult fungi, yeast, and oomycete plant pathogens. To ensure accurate and specific results, the kit's software compares your samples to a database of D2 sequences from over 1,000 validated type species.

The MicroSEQ® ID System: A Complete, Integrated Solution
Streamlining every step in microbial identification, Applied Biosystems MicroSEQ® System combines the advantages of MicroSEQ® ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ® bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Target Organism Class: Fungi
Target Gene: D2 region of rRNA gene
Validated Application: Polymerase Chain Reaction
Detection Method: Gel Electrophoresis
For Use With (Equipment): 9800 Fast Thermal Cycler, Veriti® Thermal Cycler
Sample Type (Specific): DNA (Genomic)
Technique: SBT (Sequence-Based Typing)
Library Type: Customizable, Fungal Library (> 1100 species)
Analysis Software: MicroSEQ® ID Analysis Software
Product Line: MicroSEQ™
Product Size: 1 kit
Shipping Condition: Dry Ice
Target Molecule: DNA
Workflow Step: PCR Amplification

Contents & storage

The kit contains reagents sufficient for 50 PCR reactions:
PCR Master Mix
Positive Control DNA
Negative Control (water)
After first use, store the kit in a PCR clean room at 2 to 6°C

Documents

Manuals & protocols