7900HT Fast Real-Time PCR System with Fast 96-Well Block Module

Catalog number: 4351405

Applied Biosystems™  Related applications: Genotyping & Genomic Profiling | Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 7900HT Fast Real-Time PCR System is the only real-time quantitative PCR system that combines 96- and 384-well plate compatibility and the TaqMan® Low Density Array with fully automated robotic loading-and now also offers optional Fast real-time PCR capability.

• Fast PCR option reduces run time to about 35 minutes in a standard 96-well format, or about 55 minutes in a 384-well plate.
• Continuous wavelength detection from 500-660 nm allows the use of multiple fluorophores in a single reaction.
• Measure gene expression levels, detect and quantitate pathogens, perform allelic discrimination (SNP genotyping) assays as well as score the presence of gene sequences.
• 96- or 384-well plate compatibility (including the TaqMan® Low Density Array).
• Optional Enterprise edition software provides data analysis tools that support 21 CFR Part 11 guidelines providing data integrity and security.
• Hands-free plate-loading and unloading provides true walkaway automation allowing you to increase your lab's productivity.
• Proven assay development guidelines save time and money

High Throughput
The 7900HT System is a high-throughput real-time PCR system that detects and quantitates nucleic acid sequences. An Automation Accessory combined with 384-well plate capability make the 7900HT system ideally suited to meet the high-throughput requirements of today's drug discovery process. Key applications include gene expression quantitation and the detection of single nucleotide polymorphisms (SNPs) using the fluorogenic 5' nuclease assay.

Flexibility
The 7900HT system is a versatile research tool that can accommodate any real-time PCR need. User-interchangeable thermal cycling block formats let you select the format that’s right for your project, using industry-standard 96- and 384-well formats, as well as a novel 384-well TaqMan® Low Density Array and a new Fast 96-well block that reduces run times from 2 hours to about 30 minutes. An easy automation upgrade path lets you add features to meet throughput demands. A Fast PCR option reduces run times from 2 hours to about 30 minutes.

Powerful Software
Using two automation software tools, Plate Utility and Automation Controller, most of the gene expression and SNP genotyping workflow is fully automated for high-throughput applications and requires minimal user intervention. You can now process 5,000 x 384 well plates (384 markers), which equates to 1.92 million genotypes. Using Relative Quantification (RQ) Manager, you can also process 200 x 384-well plates (96 detectors with quadruplicate data points⁄multiplexed endogenous control), which equates to 153,600 data points. Fully integrated into one complete Enterprise system, SNP Manager and RQ Manager eliminate data analysis bottlenecks in high-throughput SNP and gene expression research by allowing significantly more plates to be analyzed simultaneously. This reduces hands-on analysis time and simplifies the data analysis workflow.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: 96-well plate
Optics: No filters (CCD acts as spectrograph with continuous detection 500 -650 nm), Extended-life 488 nm argon-ion laser excitation source
Weight: 82 kg (180 lbs) without automation accessory, 114 kg (250 lbs) with automation accessory
Capacity: Up to 36 x 96-well plates with optional automation accessory, Up to 84 x 384-well plates (with optional automation accessory and 384-well block)
Run Time: 33 minutes (96-well in fast mode)
Dimensions: 72 cm (W) x 84 cm (D) x 64 cm (H) (with drawer open)
Throughput: ≥10,000 endpoint PCR rxns per hour (384-well block and automation accessory), ≥5,000 real-time PCR rxns per day (384-well block and automation accessory)
Sensitivity: Down to 10 copies
Block Format: Interchangeable
Product Size: 1 instrument
Dynamic Range: Up to 9 logs of linear dynamic range
Calibrated Dye: SYBR® Green I, TAMRA™, FAM™, JOE™, VIC™, TET™, NED™, ROX™
Reaction Speed: Fast
Detection Method: Primer-Probe Detection, SYBR
Sample Ramp Rate: Standard mode: ± 1.6°C⁄sec, Fast mode: ± 3.0°C⁄sec
Temperature Accuracy: ±0.25°C of display temperature
Passive Reference Dye: ROX (Separate Tube), ROX (Pre-mixed), No ROX, Other Passive Reference Dye
Reaction Volume Range: 10-30 µl
Temperature Uniformity: ±0.5°C
Thermal Cycling System: Peltier-Based System
For Use With (Equipment): 7900HT Fast System
Temperature Range (Metric): 4-100°C
High Throughput Compatibility: High Throughput-Compatible, Multiplexing

Contents & storage

1 Instrument with Fast 96-Well Block Module

Documents

Manuals & protocols