Applied Biosystems™

Precision Plate Holder for Plates, for 7500 Fast System

Catalog number:  4359652

 Related applications: Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Precision plate holder for 7500 fast system.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 7500 Fast System
Plate Compatibility: 96-Well Plates
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Holder

Contents & storage

7500 Fast Precision Plate Holder

Documents

Manuals & protocols