Applied Biosystems™

MicroAmp™ Optical Adhesive Film

Catalog number:  4360954

 Related applications: Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Applied Biosystems® MicroAmp® Optical Adhesive Film reduces the chance of well-to-well contamination and sample evaporation when applied to a microplate.

Features:

• Pressure-sensitive adhesive provides a tight seal over every well
• Will not interfere with sample reading
• Press-N-Seal: Align properly to the plate, then press to seal
• Easy to apply-will not stick to your glove

Prevent Loss of Precious Samples
Use this optically-clear adhesive film to seal the samples into the wells of a microplate. This will reduce the possibility of cross-contamination between sample wells and help ensure consistent Real-Time PCR data.

Easy to Use
Simply peel the backing away from the adhesive film and apply firmly across the top of your microplate. Stack microplates and read samples as you would normally except with less chance of sample contamination. Designed to fit on MicroAmp® 96-well and 384-well plates.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Optical
For Use With (Equipment): 7000 System, 7300 System, 7500 Fast Dx System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp™ 9700, ProFlex™ PCR System, QuantStudio™, QuantStudio™ Dx, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti® Dx Fast Thermal Cycler, Veriti® Dx Thermal Cycler, Veriti® Fast Thermal Cycler, Veriti® Thermal Cycler, ViiA™ 7 Dx Fast System, ViiA™ 7 Dx System, ViiA™ 7 Fast System, ViiA™ 7 System
Plate Compatibility: 384-Well Plates, 96-Well Plates
Product Line: MicroAmp®
Product Size: 25 covers
Shipping Condition: Room Temperature
Type: Film, Seal, or Applicator

Contents & storage

Includes 25 Optical Adhesive Covers. Store at room temperature.

Documents

Manuals & protocols

Recommended products