Applied Biosystems™

Precision Plate Holder for 7300 & 7500 Systems

Catalog number:  4367032

 Related applications: Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This precision plate holder is for use with Applied Biosystems® 7300 real-time PCR systems (SN 273000878 and above), as well as 7500 real-time PCR systems (SN 275001330 and above). Customers with earlier serial numbers should contact Product Support.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 7300 System, 7500 System
Plate Compatibility: 96-Well Plates
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Holder

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products