This product is now part of a food safety range that includes Thermo Scientific™ culture media, molecular solutions, and other products for your complete microbiological workflow. In US, Canada and Europe, order this product from Thermo Fisher Scientific microbiology. >> Order now
Applied Biosystems™

TaqMan™ Staphylococcus aureus Detection Kit

Catalog number:  4368606

 Related applications: Diagnostic Testing | Food & Beverage

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The TaqMan® Staphylococcus aureus Detection Kit provides a simple, reliable, and rapid procedure for detecting the presence of Staphylococcus aureus. The assay utilizes the polymerase chain reaction (PCR) to amplify a target unique to that microorganism and TaqMan® probes that identify the specific organism.

• Highly selective identification of S. aureus in a wide variety of samples
• Verified performance
• Ready-to-use convenience
• Minimal risk of contamination

3 Easy Steps
This kit has 3 easy steps: Enrich samples, prepare samples, and run samples. Grow the specific pathogen from a selected sample using a standard enrichment procedure in the Enrichment step. In the Sample Preparation step isolate the DNA for PCR, and prepare, mix and pipet the reactions. The final step is Running a Plate on an Applied Biosystems® SDS or Real-Time PCR System to analyze your sample.

A Complete Solution
We offer a full line of TaqMan® Pathogen Detection kits based on universal assay conditions, enabling you to run any combination of these assays together in the same sample run. In combination with our real-time PCR systems, the TaqMan® Staphylococcus aureus. Detection Kit offers a fully integrated, single-vendor solution to identify Staphylococcus aureus..

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Target Organism Class: S. aureus
Validated Application: Real-Time PCR
Detection Method: Primer-Probe Detection
PCR Method: qPCR
For Use With (Equipment): 7300 System, 7500 System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
Sample Type (General): Bacteria, Food & Environmental Samples
Sample Type (Specific): Any Swab Type, Food, Water Samples
Hot Start: Built-In Hot Start
Passive Reference Dye: ROX (Pre-mixed)
Reaction Speed: Standard
Primer-Probe Compatibility: TaqMan®
Polymerase: AmpliTaq Gold® DNA Polymerase
Number of Reactions: 100 Tests
Product Line: TaqMan™
Product Size: 1 kit
Shipping Condition: Dry Ice

Contents & storage

This kit contains Target Assay Mix (store at -20°C in the dark), a Negative Control (store at -2 to 8°C), and 2X Environmental Master Mix (store at -2 to 8°C).

There are enough reagents for 100 assays. These reagents are stable for one year after shipment.

Documents

Manuals & protocols