Fast MicroSEQ™ 500 16S rDNA PCR Kit, with quick reference card & protocol

Catalog number: 4370653

Applied Biosystems™  Related applications: Microbial Identification & Detection | Microbial Protein Production

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Fast MicroSEQ® 500 16S rDNA PCR kit utilizes new Fast technology and provides bacterial identifications from colony to results in as little as 5 hours using the Veriti® 96-well Thermal Cycler. This kit is also compatable with the GeneAmp® PCR System 9700. The PCR-amplified products that are generated using this kit can be sequenced using the MicroSEQ® 500 16S rDNA Seqencing Kit. The two kits together permit the amplification and sequencing of 500 base pairs of the 16S rRNA gene. Thses sequences can then be compared to the MicroSEQ® ID Analysis Software 16S rDNA 500 Library to obtain an identity match. This kit does include a protocol and a Quick Reference Card.

• Sequence-based measurement ensures you get reproducible results in under 24 hours.
• Growth requirement-independent measurement enables you to identify bacteria using objective criteria.
• Universal domain-level primers eliminate up-front optimization.
• MicroSEQ® ID Analysis Software ensures accurate species identification.
• Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
• Requires minimal training-same work flow as the MicroSEQ® D2 LSU fungal kit.

The MicroSEQ® ID System: A Complete, Integrated Solution
Streamlining every step in microbial identification, Applied Biosystems® MicroSEQ® System combines the advantages of MicroSEQ®: ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ® bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Target Organism Class: Bacteria
Target Gene: 1st 500 bp of 16S rRNA gene
Validated Application: Polymerase Chain Reaction
Detection Method: Gel Electrophoresis
For Use With (Equipment): Veriti® Thermal Cycler
Sample Type (Specific): DNA (Genomic)
Technique: SBT (Sequence-Based Typing)
Library Type: Bacterial Library (> 2000 species), Customizable
Analysis Software: MicroSEQ® ID Analysis Software
Product Line: MicroSEQ™
Product Size: 1 kit
Target Molecule: DNA
Target Specificity: Target-Specific
Workflow Step: PCR Amplification

Contents & storage

Each kit contains enough PCR Master Mix to perform 50 PCR amplifications, 5 negative control assays, and 5 positive control assays. One tube of positive control E. coli DNA, and one of water (for use as a negative control) are also included. Store at -15 to -25°C upon reciept. After first use, store at 2-6 °C in a PCR Clean Room.

Protocol and Quick Reference Guide are included

Documents

Manuals & protocols