Applied Biosystems™

AgPath-ID™ One-Step RT-PCR Reagents

Catalog number:  4387424

 Related applications: Animal Health | Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

AgPath-ID™ One-Step RT-PCR Reagents are designed for sensitive, robust amplification of RNA targets using a rapid single-tube TaqMan® real-time reverse transcription PCR (RT-PCR) strategy.
• Recommended for RNA pathogen amplification
• Consistent amplification of RNA targets with high specificity and sensitivity
• Optimized to work with your target-specific primers and probes
• Contains ROX for quantitative fluorescent signal normalization

AgPath-ID™ One-Step RT-PCR Reagents are configured for fast and simple reaction setup. The reactions are assembled in a single tube, minimizing sample handling errors and expediting set-up time. Once assembled, results are available in approximately one hour. The 25X RT-PCR Enzyme Mix included in the kit contains highly efficient ArrayScript™ Reverse Transcriptase, a mutant MMLV RT that produces high cDNA yields, and AmpliTaq Gold® polymerase, the preferred hot-start DNA polymerase for specific target amplification. The 2X RT-PCR Buffer has been optimized for efficient, robust reverse transcription and PCR includes the passive reference dye, ROX™ Dye, for quantitative fluorescent signal normalization. A Detection Enhancer is also provided as an optional reagent for amplification of templates with a high GC content or persistent secondary structure.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

PCR Method: 1-Step qRT-PCR
Reverse Transcriptase: ArrayScript™
Polymerase: AmpliTaq Gold® DNA Polymerase
Detection Method: Primer-Probe Detection
Primer-Probe Compatibility: TaqMan®
Reaction Speed: Standard
For Use With (Equipment): 7500 System
Sample Type (General): RNA
Passive Reference Dye: ROX (Pre-mixed)
Comments: Works on all AB platforms, but is optimized for the 7500.
Product Line: AgPath-ID™, Ambion™
Product Size: 500 reactions
Shipping Condition: Dry Ice

Contents & storage

Sufficient for 500 reactions of 25 µL, contains:
• 7mL 2X RT-PCR Buffer
• 550 µL 25X RT-PCR Enzyme Mix
• 1.2 mL Detection Enhancer
• 25 mL Nuclease-free Water.

Documents

Manuals & protocols

Recommended products