MagMAX™ Express-96 Standard Magnetic Head

Catalog number: 4388434

Applied Biosystems™  Related applications: Animal Health Sample Preparation | Automated Nucleic Acid Purification | RNA Extraction | Viral RNA⁄DNA Purification

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Applied Biosystems® Replacement head for the MagMAX™ Express-96 Standard Magnetic Particle Processors (SKUs #4400076 and #4400078). 1 head is included.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type: Magnetic Head
Product Size: 1 each
Shipping Condition: Room Temperature
For Use With (Equipment): MagMAX™ Express-96 Standard Magnetic Particle Processor

Contents & storage

Store at room temperature.

Documents

Manuals & protocols