Applied Biosystems™

MagMAX™ Express-96 Deep Well Magnetic Head

Catalog number:  4388435

 Related applications: Animal Health | Automated Nucleic Acid Purification | RNA Extraction | Viral RNA⁄DNA Purification

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Applied Biosystems® Replacement head for the MagMAX™ Express-96 Deep Well Magnetic Particle Processors (SKUs #4400077 and #4400079). 1 head is included.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): MagMAX™ Express-96 Deep Well Magnetic Particle Processor
Product Line: MagMAX™
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Magnetic Head

Contents & storage

Store at room temperature.

Documents

Manuals & protocols