MagMAX™ Express Tip Combs

Catalog number: 4388452

Applied Biosystems™  Related applications: Animal Health | Automated Nucleic Acid Purification | RNA Extraction | Viral RNA⁄DNA Purification

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Applied Biosystems® Replacement combs for the MagMAX™ Express Magnetic Particle Processors (SKUs #4400074 and #4400075). 50 combs are included.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): MagMAX™ Express Magnetic Particle Processor
Product Line: Applied Biosystems™, MagMAX™
Product Size: 50 pieces
Shipping Condition: Room Temperature
Type: Tip Combs

Contents & storage

Store at room temperature.

Documents

Manuals & protocols